siRNA Details
| Gene Symbol: LOXL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC015656.2 | - | 2 | 907 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000433374.1 | - | 2 | 907 | Chr. 10 - 98252023 - 98256575 [+] on Build GRCh38 | |

LOXL4
Genomic Map