siRNA Details
| Gene Symbol: FOXN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005258046.3 | XP_005258103.1 | 9 | 3584 | ||
| XM_011525358.1 | XP_011523660.1 | 9 | 3587 | |||
| XM_011525359.1 | XP_011523661.1 | 9 | 3570 | |||
| XM_011525362.1 | XP_011523664.1 | 9 | 3468 | |||
| XM_011525367.1 | XP_011523669.1 | 7 | 3375 | |||
| XM_011525368.2 | XP_011523670.1 | 7 | 3004 | |||
| XM_011525369.2 | XP_011523671.1 | 7 | 3152 | |||
| XM_011525370.2 | XP_011523672.1 | 7 | 3149 | |||
| XM_017025228.1 | XP_016880717.1 | 9 | 3599 | |||
| XM_017025229.1 | XP_016880718.1 | 9 | 3324 | |||
| XM_017025230.1 | XP_016880719.1 | 8 | 3078 | |||
| GenBank mRNA | AF086453.1 | - | 1 | 531 | ||

FOXN1
Genomic Map