siRNA Details
| Gene Symbol: LOC284561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC034299.1 | - | 2 | 196 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI830630.1 | - | 2 | 195 | Chr. 1 - 148295180 - 148296503 [+] on Build GRCh38 | |

LOC284561
Genomic Map