siRNA Details
Gene Symbol: ELFN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055475.1 | - | 3 | 1755 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000430883.5 | - | 3 | 1755 | Chr. 22 - 37339583 - 37385283 [-] on Build GRCh38 |
