siRNA Details
| Gene Symbol: HEXDC-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK129961.1 | - | 1 | 1126 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HEXDC-IT1| Gene Symbol: HEXDC-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK129961.1 | - | 1 | 1126 | ||