siRNA Details
| Gene Symbol: LOC101927358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121182.1 | 1 | 101 | |||
| GenBank mRNA | BC021182.1 | - | 1 | 106 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG720148.1 | - | 1 | 106 | Chr. 9 - 74371531 - 74383530 [+] on Build GRCh38 | |

LOC101927358
Genomic Map