siRNA Details
| Gene Symbol: LINC00561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC034981.2 | - | 1 | 2586 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00561| Gene Symbol: LINC00561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC034981.2 | - | 1 | 2586 | ||