siRNA Details
| Gene Symbol: AJAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011541786.2 | XP_011540088.1 | 7 | 12002 | ||
| GenBank mRNA | BC037321.2 | - | 1 | 2812 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AJAP1| Gene Symbol: AJAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011541786.2 | XP_011540088.1 | 7 | 12002 | ||
| GenBank mRNA | BC037321.2 | - | 1 | 2812 | ||