siRNA Details
| Gene Symbol: LINC01297 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY338954.1 | - | 3 | 4687 | ||
| Gene Symbol: LINC01087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108087.1 | 1 | 1835 | |||
| GenBank mRNA | BC036114.1 | - | 1 | 1832 | ||

LINC01297
Genomic Map