siRNA Details
| Gene Symbol: AGO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017629.3 | NP_060099.2 | 18 | 6674 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGO4| Gene Symbol: AGO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017629.3 | NP_060099.2 | 18 | 6674 | ||