siRNA Details
| Gene Symbol: AGO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017629.3 | NP_060099.2 | 18 | 6098 | ||
| XM_005270578.3 | XP_005270635.1 | 18 | 5983 | |||
| XM_005270579.3 | XP_005270636.1 | 17 | 5791 | |||
| XM_011540883.2 | XP_011539185.1 | 15 | 5587 | |||
| XM_011540884.2 | XP_011539186.1 | 15 | 5450 | |||

AGO4
Genomic Map