siRNA Details
| Gene Symbol: AGO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017629.3 | NP_060099.2 | 18 | 5443 | ||
| XM_005270578.3 | XP_005270635.1 | 18 | 5328 | |||
| XM_005270579.3 | XP_005270636.1 | 17 | 5136 | |||
| XM_011540883.2 | XP_011539185.1 | 15 | 4932 | |||
| XM_011540884.2 | XP_011539186.1 | 15 | 4795 | |||
| GenBank mRNA | AB046787.1 | - | 18 | 5443 | ||
| BC152450.1 | - | 18 | 5443 | 8862582 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR598503.1 | - | 1 | 191 | Chr. 1 - 35856082 - 35857675 [+] on Build GRCh38 | |

AGO4
Genomic Map