siRNA Details
| Gene Symbol: TTLL7-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC042527.1 | - | 2 | 1033 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTLL7-IT1| Gene Symbol: TTLL7-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC042527.1 | - | 2 | 1033 | ||