siRNA Details
| Gene Symbol: DYRK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017020032.1 | XP_016875521.1 | 2 | 7190 | ||
| GenBank mRNA | AK024870.1 | - | 1 | 1081 | ||
| AL832817.1 | - | 1 | 2608 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DYRK2| Gene Symbol: DYRK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017020032.1 | XP_016875521.1 | 2 | 7190 | ||
| GenBank mRNA | AK024870.1 | - | 1 | 1081 | ||
| AL832817.1 | - | 1 | 2608 | |||