siRNA Details
| Gene Symbol: LINC00208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040035.1 | 3 | 1709 | |||
| GenBank mRNA | AJ291678.1 | - | 3 | 1709 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00208| Gene Symbol: LINC00208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040035.1 | 3 | 1709 | |||
| GenBank mRNA | AJ291678.1 | - | 3 | 1709 | ||