siRNA Details
| Gene Symbol: ZNF585A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288800.1 | NP_001275729.1 | 5 | 4395 | ||
| NM_152655.3 | NP_689868.1 | 6 | 4497 | |||
| NM_199126.2 | NP_954577.1 | 6 | 4437 | |||
| GenBank mRNA | AK123296.1 | - | 1 | 373 | ||

ZNF585A
Genomic Map