siRNA Details
| Gene Symbol: LOC145694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132970.1 | 3 | 1767 | |||
| NR_132971.1 | 2 | 1712 | ||||
| GenBank mRNA | AK056793.1 | - | 3 | 1767 | ||
| BX097197.1 | - | 1 | 451 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC145694| Gene Symbol: LOC145694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132970.1 | 3 | 1767 | |||
| NR_132971.1 | 2 | 1712 | ||||
| GenBank mRNA | AK056793.1 | - | 3 | 1767 | ||
| BX097197.1 | - | 1 | 451 | |||