siRNA Details
| Gene Symbol: LOC145694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132970.1 | 3 | 1166 | |||
| NR_132971.1 | 2 | 1111 | ||||
| GenBank mRNA | AK056793.1 | - | 3 | 1166 | ||
| CN360125.1 | - | 2 | 285 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000558617.1 | - | 3 | 1166 | Chr. 15 - 69396904 - 69415029 [-] on Build GRCh38 | |

LOC145694
Genomic Map