siRNA Details
Gene Symbol: LOC145694 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132970.1 | 3 | 1323 | |||
NR_132971.1 | 2 | 1268 | ||||
GenBank mRNA | AK056793.1 | - | 3 | 1323 | ||
CN360125.1 | - | 2 | 442 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000558617.1 | - | 3 | 1323 | Chr. 15 - 69396904 - 69415029 [-] on Build GRCh38 |
