siRNA Details
| Gene Symbol: LOC145694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132970.1 | 3 | 1118 | |||
| NR_132971.1 | 2 | 1063 | ||||
| GenBank mRNA | AK056793.1 | - | 3 | 1118 | ||
| CN360125.1 | - | 2 | 237 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC145694| Gene Symbol: LOC145694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132970.1 | 3 | 1118 | |||
| NR_132971.1 | 2 | 1063 | ||||
| GenBank mRNA | AK056793.1 | - | 3 | 1118 | ||
| CN360125.1 | - | 2 | 237 | |||