siRNA Details
| Gene Symbol: TMEM72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123376.2 | NP_001116848.1 | 5 | 2308 | ||
| XM_011540069.2 | XP_011538371.1 | 5 | 5110 | |||
| XM_011540070.2 | XP_011538372.1 | 5 | 5056 | |||
| XM_011540071.2 | XP_011538373.1 | 6 | 2417 | |||
| GenBank mRNA | AB235418.1 | - | 5 | 2263 | ||
| BC029135.1 | - | 1 | 112 | |||

TMEM72
Genomic Map