siRNA Details
| Gene Symbol: LOC340107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037896.1 | 1 | 216 | |||
| GenBank mRNA | BC040315.1 | - | 1 | 216 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG773452.1 | - | 1 | 220 | Chr. 5 - 24836865 - 24840585 [-] on Build GRCh38 | |

LOC340107
Genomic Map