siRNA Details
| Gene Symbol: MID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY539962.1 | - | 1 | 18 | ||
| AY539963.1 | - | 1 | 57 | |||
| AY539964.1 | - | 1 | 18 | |||
| AY539965.1 | - | 1 | 18 | |||
| AY539966.1 | - | 1 | 15 | |||
| AY539967.1 | - | 1 | 18 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MID1| Gene Symbol: MID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY539962.1 | - | 1 | 18 | ||
| AY539963.1 | - | 1 | 57 | |||
| AY539964.1 | - | 1 | 18 | |||
| AY539965.1 | - | 1 | 18 | |||
| AY539966.1 | - | 1 | 15 | |||
| AY539967.1 | - | 1 | 18 | |||