siRNA Details
| Gene Symbol: KDM4A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033827.1 | 5 | 849 | |||
| GenBank mRNA | BC073819.1 | - | 5 | 643 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM918225.1 | - | 5 | 641 | Chr. 1 - 43700119 - 43707133 [-] on Build GRCh38 | |

KDM4A-AS1
Genomic Map