siRNA Details
| Gene Symbol: PHLPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK001854.1 | - | 9 | 1627 | ||
| BC035267.1 | - | 15 | 2648 | |||
| BX648394.1 | - | 2 | 1946 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PHLPP2| Gene Symbol: PHLPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK001854.1 | - | 9 | 1627 | ||
| BC035267.1 | - | 15 | 2648 | |||
| BX648394.1 | - | 2 | 1946 | |||