siRNA Details
| Gene Symbol: IRAK1BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010844.3 | NP_001010844.1 | 4 | 3008 | ||
| GenBank mRNA | AK096861.1 | - | 1 | 1680 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IRAK1BP1| Gene Symbol: IRAK1BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010844.3 | NP_001010844.1 | 4 | 3008 | ||
| GenBank mRNA | AK096861.1 | - | 1 | 1680 | ||