siRNA Details
| Gene Symbol: IRAK1BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010844.3 | NP_001010844.1 | 4 | 3219 | ||
| GenBank mRNA | AK096861.1 | - | 1 | 1891 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX109533.1 | - | 1 | 565 | Chr. 6 - 78900172 - 78900966 [+] on Build GRCh38 | |

IRAK1BP1
Genomic Map