siRNA Details
| Gene Symbol: LOC101927583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120450.1 | 4 | 2363 | |||
| GenBank mRNA | BC050035.1 | - | 4 | 2363 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927583| Gene Symbol: LOC101927583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120450.1 | 4 | 2363 | |||
| GenBank mRNA | BC050035.1 | - | 4 | 2363 | ||