siRNA Details
| Gene Symbol: LOC100505609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132416.1 | 2 | 352 | |||
| GenBank mRNA | BC041442.1 | - | 2 | 351 | ||
| BI830672.1 | - | 2 | 353 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100505609| Gene Symbol: LOC100505609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132416.1 | 2 | 352 | |||
| GenBank mRNA | BC041442.1 | - | 2 | 351 | ||
| BI830672.1 | - | 2 | 353 | |||