siRNA Details
| Gene Symbol: MIR194-2HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133638.1 | 2 | 2450 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK130408.1 | - | 11 | Chr. 11 - 64889563 - 64890767 [-] on Build GRCh38 | ||

MIR194-2HG
Genomic Map