siRNA Details
| Gene Symbol: TOR1AIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022347.3 | NP_071742.1 | 3 | 3598 | ||
| XM_017000477.1 | XP_016855966.1 | 2 | 4271 | |||
| XM_017000478.1 | XP_016855967.1 | 2 | 3604 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TOR1AIP2| Gene Symbol: TOR1AIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022347.3 | NP_071742.1 | 3 | 3598 | ||
| XM_017000477.1 | XP_016855966.1 | 2 | 4271 | |||
| XM_017000478.1 | XP_016855967.1 | 2 | 3604 | |||