siRNA Details
| Gene Symbol: LOC101927272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110908.1 | 3 | 643 | |||
| GenBank mRNA | AF143871.1 | - | 3 | 652 | ||
| AL161979.1 | - | 2 | 4809 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927272| Gene Symbol: LOC101927272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110908.1 | 3 | 643 | |||
| GenBank mRNA | AF143871.1 | - | 3 | 652 | ||
| AL161979.1 | - | 2 | 4809 | |||