siRNA Details
| Gene Symbol: RIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017027893.1 | XP_016883382.1 | 9 | 4441 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK130346.1 | - | 1 | 607 | Chr. 20 - 19977350 - 19980044 [+] on Build GRCh38 | |

RIN2
Genomic Map