siRNA Details
Gene Symbol: OPHN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK309696.1 | - | 3 | 1054 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK130340.1 | - | 1 | 1090 | Chr. X - 68380510 - 68383984 [-] on Build GRCh38 |
