siRNA Details
| Gene Symbol: OPHN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK309696.1 | - | 3 | 996 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK130340.1 | - | 1 | 1032 | Chr. X - 68380510 - 68383984 [-] on Build GRCh38 | |

OPHN1
Genomic Map