siRNA Details
| Gene Symbol: CAPN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK125713.1 | - | 4 | 1280 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK094707.1 | - | 1 | 1258 | Chr. 19 - 38733121 - 38735672 [-] on Build GRCh38 | |

CAPN12
Genomic Map