siRNA Details
| Gene Symbol: LOC105377468 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL832220.1 | - | 1 | 3529 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105377468| Gene Symbol: LOC105377468 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL832220.1 | - | 1 | 3529 | ||