siRNA Details
| Gene Symbol: PDCD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK308872.1 | - | 4 | 640 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK124667.1 | - | 1 | 1111 | Chr. 5 - 310457 - 312809 [+] on Build GRCh38 | |

PDCD6
Genomic Map