siRNA Details
Gene Symbol: LOC284454 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036515.1 | 1 | 55 | |||
GenBank mRNA | AL832183.1 | - | 1 | 54 | ||
BX640708.1 | - | 1 | 55 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY171639.1 | - | 1 | 54 | Chr. 19 - 13835793 - 13836288 [-] on Build GRCh38 |
