siRNA Details
Gene Symbol: LOC284454 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036515.1 | 1 | 1575 | |||
GenBank mRNA | AL832183.1 | - | 1 | 1571 | ||
BX640708.1 | - | 1 | 1575 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC284454 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036515.1 | 1 | 1575 | |||
GenBank mRNA | AL832183.1 | - | 1 | 1571 | ||
BX640708.1 | - | 1 | 1575 |