siRNA Details
Gene Symbol: LAMP5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109957.1 | 1 | 33 | |||
GenBank mRNA | AK096264.1 | - | 1 | 33 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA696611.1 | - | 1 | 33 | Chr. 20 - 9507206 - 9514998 [-] on Build GRCh38 |
