siRNA Details
| Gene Symbol: LAMP5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109957.1 | 5 | 1863 | |||
| GenBank mRNA | AK096264.1 | - | 5 | 1863 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LAMP5-AS1| Gene Symbol: LAMP5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109957.1 | 5 | 1863 | |||
| GenBank mRNA | AK096264.1 | - | 5 | 1863 | ||