siRNA Details
Gene Symbol: LAMP5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109957.1 | 5 | 1378 | |||
GenBank mRNA | AK096264.1 | - | 5 | 1378 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000443469.1 | - | 5 | 1378 | Chr. 20 - 9505180 - 9514998 [-] on Build GRCh38 |
