siRNA Details
| Gene Symbol: DNAJB14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031723.3 | NP_001026893.1 | 8 | 5408 | ||
| NM_001278310.1 | NP_001265239.1 | 7 | 5236 | |||
| GenBank mRNA | AK095074.1 | - | 1 | 1492 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNAJB14| Gene Symbol: DNAJB14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031723.3 | NP_001026893.1 | 8 | 5408 | ||
| NM_001278310.1 | NP_001265239.1 | 7 | 5236 | |||
| GenBank mRNA | AK095074.1 | - | 1 | 1492 | ||