siRNA Details
| Gene Symbol: DCAF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK022091.1 | - | 127 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DCAF8| Gene Symbol: DCAF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK022091.1 | - | 127 | |||