siRNA Details
| Gene Symbol: SAMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174156.1 | NP_001167627.1 | 6 | 3137 | ||
| NM_144660.2 | NP_653261.1 | 5 | 3364 | |||
| XM_005269541.4 | XP_005269598.1 | 6 | 3328 | |||
| XM_011539311.1 | XP_011537613.1 | 6 | 3256 | |||
| XM_011539312.2 | XP_011537614.1 | 6 | 3186 | |||
| XM_017015738.1 | XP_016871227.1 | 6 | 3193 | |||
| XM_017015739.1 | XP_016871228.1 | 6 | 3221 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK055455.1 | - | 169 | Chr. 14 - 67770973 - 67772857 [-] on Build GRCh38 | ||

SAMD8
Genomic Map