siRNA Details
| Gene Symbol: FRMD6-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037676.1 | 1 | 1677 | |||
| GenBank mRNA | AK022437.1 | - | 1 | 1678 | ||
| BX104826.1 | - | 1 | 35 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | N34909.1 | - | 1 | 215 | Chr. 14 - 51649983 - 51650282 [-] on Build GRCh38 | |

FRMD6-AS1
Genomic Map