siRNA Details
| Gene Symbol: FAM120B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286379.1 | NP_001273308.1 | 11 | 5061 | ||
| NM_001286380.1 | NP_001273309.1 | 11 | 4824 | |||
| NM_001286381.1 | NP_001273310.1 | 9 | 2876 | |||
| NM_032448.2 | NP_115824.1 | 11 | 4812 | |||
| XM_017011357.1 | XP_016866846.1 | 9 | 3032 | |||
| GenBank mRNA | AJ420589.1 | - | 1 | 1139 | ||

FAM120B
Genomic Map