siRNA Details
| Gene Symbol: MIR194-2HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133638.1 | 2 | 209 | |||
| GenBank mRNA | AK092802.1 | - | 2 | 209 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA922338.1 | - | 2 | 209 | Chr. 11 - 64892641 - 64893449 [-] on Build GRCh38 | |

MIR194-2HG
Genomic Map