siRNA Details
| Gene Symbol: MIR194-2HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133638.1 | 2 | 1448 | |||
| GenBank mRNA | AK092802.1 | - | 2 | 1448 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR194-2HG| Gene Symbol: MIR194-2HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133638.1 | 2 | 1448 | |||
| GenBank mRNA | AK092802.1 | - | 2 | 1448 | ||